Learn More
Abnova™ Human MEF2A Full-length ORF (AAH13437, 1 a.a. - 499 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression. The myogenic basic helix-loop-helix proteins, including myoD (MIM 159970), myogenin (MIM 159980), MYF5 (MIM 159990), and MRF4 (MIM 159991) are one class of identified factors. A second family of DNA binding regulatory proteins is the myocyte-specific enhancer factor-2 (MEF2) family. Each of these proteins binds to the MEF2 target DNA sequence present in the regulatory regions of many, if not all, muscle-specific genes. The MEF2 genes are members of the MADS gene family (named for the yeast mating type-specific transcription factor MCM1, the plant homeotic genes 'agamous' and 'deficiens' and the human serum response factor SRF (MIM 600589)), a family that also includes several homeotic genes and other transcription factors, all of which share a conserved DNA-binding domain.[supplied by OMIM]
Spezifikation
Spezifikation
Zugriffsnummer | AAH13437 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 4205 |
Molekulargewicht | 80.41kDa |
Name | MEF2A (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 ug |
Immunogen | MGRKKIQITRIMDERNRQVTFTKRKFGLMKKAYELSVLCDCEIALIIFNSSNKLFQYASTDMDKVLLKYTEYNEPHESRTNSDIVEALNKKEHRGCDSPDPDTSYVLTPHTEEKYKKINEEFDNMMRNHKIAPGLPPQNFSMSVTVPVTSPNALSYTNPGSSLVSPSLAASSTLTDSSMLSPPQTTLHRNVSPGAPQRPPSTGNAGGMLSTTDLTVPNGAGSSPVGNGFVNSRASPNLIGATGANSLGKVMPTKSPPPPGGGNLGMNSRRPDLRVVIPPSSKGMMPPLNTQRISSSQATQPLATPVVSVTTPSLPPQGLVYSAMPTAYNTDYSLTSADLSALQGFNSPGMLSLGQVSAWQQHHLGQAALSSLVAGGQLSQGSNLSINTNQNISIKSEPISPPRDRMTPSGFQQQQQQQQQQQPPPPPQPQPQPPQPQPRQEMGRSPVDSLSSSSSSYDGSDREDPRGDFHSPIVLGRPPNTEDRESPSVKRMRMDAWVT |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.