missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MEA1 Partial ORF (NP_055438.1, 121 a.a. - 185 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004201-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: GATALNNHSSIPMDPEHVELVKRTMAGVSLPAPGVPAWAREISDAQWEDVVQKALQARQASPAWKSpezifikation
NP_055438.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GATALNNHSSIPMDPEHVELVKRTMAGVSLPAPGVPAWAREISDAQWEDVVQKALQARQASPAWK | |
RUO | |
MEA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4201 | |
MEA1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HYS/MEA | |
MEA1 | |
Recombinant | |
wheat germ expression system |