Learn More
Abnova™ Human MCM4 Full-length ORF (AAH31061.1, 1 a.a. - 863 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004173-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]
Sequence: MSSPASTPSRRGSRRGRATPAQTPRSEDARSSPSQRRRGEDSTSTGELQPMPTSPGVDLQSPAAQDVLFSSPPQMHSSAIPLDFDVSSPLTYGTPSSRVEGTPRSGVRGTPVRQRPDLGSAQKGLQVDLQSDGAAAEDIVASEQSLGQKLVIWGTDVNVAACKENFQRFLQRFIDPLAKEEENVGIDITEPLYMQRLGEINVIGEPFLNVNCEHIKSFDKNLYRQLISYPQEVIPTFDMAVNEIFFDRYPDSILEHQIQVRPFNALKTKNMRNLNPEDIDQLITISGMVIRTSQLIPEMQEAFFQCQVCAHTTRVEMDRGRIAEPSVCGRCHTTHSMALIHNRSLFSDKQMIKLQESPEDMPAGQTPHTVILFAHNDLVDKVQPGDRVNVTGIYRAVPIRVNPRVSNVKSVYKTHIDVIHYRKTDAKRLHGLDEEAEQKLFSEKRVELLKELSRKPDIYERLASALAPSIYEHEDIKKGILLQLFGGTRKDFSHTGRGKFRAEINILLCGDPGTSKSQLLQYVYNLVPRGQYTSGKGSSAVGLTAYVMKDPETRQLVLQTGALVLSDNGICCIDEFDKMNESTRSVLHEVMEQQTLSIAKAGIICQLNARTSVLAAANPIESQWNPKKTTIENIQLPHTLLSRFDLIFLMLDPQDEAYDRRLAHHLVALYYQSEEQAEEELLDMAVLKDYIAYAHSTIMPRLSEEASQALIEAYVDMRKIGSSRGMVSAYPRQLESLIRLAEAHAKVRLSNKVEAIDVEEAKRLHREALKQSATDPRTGIVDISILTTGMSATSRKRKEELAEALKKLILSKGKTPALKYQQLFEDIRGQSDIAITKDMFEEALRALADDDFLTVTGKTVRLLSpezifikation
AAH31061.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
123kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDC21/CDC54/MGC33310/P1-CDC21/hCdc21 | |
MCM4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4173 | |
MCM4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSPASTPSRRGSRRGRATPAQTPRSEDARSSPSQRRRGEDSTSTGELQPMPTSPGVDLQSPAAQDVLFSSPPQMHSSAIPLDFDVSSPLTYGTPSSRVEGTPRSGVRGTPVRQRPDLGSAQKGLQVDLQSDGAAAEDIVASEQSLGQKLVIWGTDVNVAACKENFQRFLQRFIDPLAKEEENVGIDITEPLYMQRLGEINVIGEPFLNVNCEHIKSFDKNLYRQLISYPQEVIPTFDMAVNEIFFDRYPDSILEHQIQVRPFNALKTKNMRNLNPEDIDQLITISGMVIRTSQLIPEMQEAFFQCQVCAHTTRVEMDRGRIAEPSVCGRCHTTHSMALIHNRSLFSDKQMIKLQESPEDMPAGQTPHTVILFAHNDLVDKVQPGDRVNVTGIYRAVPIRVNPRVSNVKSVYKTHIDVIHYRKTDAKRLHGLDEEAEQKLFSEKRVELLKELSRKPDIYERLASALAPSIYEHEDIKKGILLQLFGGTRKDFSHTGRGKFRAEINILLCGDPGTSKSQLLQYVYNLVPRGQYTSGKGSSAVGLTAYVMKDPETRQLVLQTGALVLSDNGICCIDEFDKMNESTRSVLHEVMEQQTLSIAKAGIICQLNARTSVLAAANPIESQWNPKKTTIENIQLPHTLLSRFDLIFLMLDPQDEAYDRRLAHHLVALYYQSEEQAEEELLDMAVLKDYIAYAHSTIMPRLSEEASQALIEAYVDMRKIGSSRGMVSAYPRQLESLIRLAEAHAKVRLSNKVEAIDVEEAKRLHREALKQSATDPRTGIVDISILTTGMSATSRKRKEELAEALKKLILSKGKTPALKYQQLFEDIRGQSDIAITKDMFEEALRALADDDFLTVTGKTVRLL | |
RUO | |
MCM4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |