Learn More
R&D Systems™ Human MBP DuoSet ELISA
Artikelnummer. 30020967
Change view
Click to view available options
Menge:
1 Kit (für 5 Platten)
Packungsgröße:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30020967 1 Kit (für 5 Platten) 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 30020967 Lieferant R&D Systems™ Lieferanten-Nr. DY422805
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs
The Human MBP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MBP.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 1 Kit (für 5 Platten)
Spezies Human
Konjugat Biotin
Bindungsziel MBP
Assay-Bereich 234.40 to 15000 pg/mL
Zur Verwendung mit (Anwendung) ELISA
Gen-ID (Entrez) 4155
Synonym Hmbpr, MGC99675, mld, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi
Inhalt und Lagerung Store the unopened product at 2°C to 8°C. Do not use past expiration date.