Learn More
Abnova™ Human MBD4 Partial ORF (NP_003916.1, 472 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008930-Q01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MBD4 may function to mediate the biological consequences of the methylation signal. In addition, MBD4 has protein sequence similarity to bacterial DNA repair enzymes and thus may have some function in DNA repair. Further, MBD4 gene mutations are detected in tumors with primary microsatellite-instability (MSI), a form of genomic instability associated with defective DNA mismatch repair, and MBD4 gene meets 4 of 5 criteria of a bona fide MIS target gene. [provided by RefSeq]
Sequence: KMAIPVLWKFLEKYPSAEVARTADWRDVSELLKPLGLYDLRAKTIVKFSDEYLTKQWKYPIELHGIGKYGNDSYRIFCVNEWKQVHPEDHKLNKYHDWLWENHEKLSLSpezifikation
NP_003916.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KMAIPVLWKFLEKYPSAEVARTADWRDVSELLKPLGLYDLRAKTIVKFSDEYLTKQWKYPIELHGIGKYGNDSYRIFCVNEWKQVHPEDHKLNKYHDWLWENHEKLSL | |
RUO | |
MBD4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8930 | |
MBD4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MED1 | |
MBD4 | |
Recombinant | |
wheat germ expression system |