missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAN1A1 Full-length ORF (AAH65827.1, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00004121-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16127601

  • 379.00 CHF / 10 Mikrogramm
Verfügbar ab: 15-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the removal of 3 distinct mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases. [provided by RefSeq]

Sequence: MPVGGLLPLFSSPAGGVLGGGLGGGGGRKGSGPAALRLTEKFVLLLVFSAFITLCFGAIFFLPDSSKLLSGVLFHSSPALQPAADHKPGPGARAEDAAEGRARRREEGAPGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKIKEEIEARKGQANCPGSSSSCEVEIQMMKHAWNNYKGYAWGLNELKPISKGGHSSSLFGNIKGATIVDALDTLFIMEMKHEFEEAKSWVEENLDFNVNAEISVFEVNIRFVGGLLSAYYLSGEEIFRKKAVELGVKLLPAFHTPSGIPWALLNMKRWMCNIPRAMEATI
Spezifikation

Spezifikation

AAH65827.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HUMM3/HUMM9/MAN9
MAN1A1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4121
MAN1A1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPVGGLLPLFSSPAGGVLGGGLGGGGGRKGSGPAALRLTEKFVLLLVFSAFITLCFGAIFFLPDSSKLLSGVLFHSSPALQPAADHKPGPGARAEDAAEGRARRREEGAPGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKIKEEIEARKGQANCPGSSSSCEVEIQMMKHAWNNYKGYAWGLNELKPISKGGHSSSLFGNIKGATIVDALDTLFIMEMKHEFEEAKSWVEENLDFNVNAEISVFEVNIRFVGGLLSAYYLSGEEIFRKKAVELGVKLLPAFHTPSGIPWALLNMKRWMCNIPRAMEATI
RUO
MAN1A1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote