missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAML2 Partial ORF (NP_115803, 796 a.a. - 894 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084441-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: RPPPDYKDQRRNVGNMQPTAQYSGGSSTISLNSNQALANPVSTHTILTPNSSLLSTSHGTRMPSLSTAVQNMGMYGNLPCNQPNTYSVTSGMNQLTQQRSpezifikation
NP_115803 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RPPPDYKDQRRNVGNMQPTAQYSGGSSTISLNSNQALANPVSTHTILTPNSSLLSTSHGTRMPSLSTAVQNMGMYGNLPCNQPNTYSVTSGMNQLTQQR | |
RUO | |
MAML2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84441 | |
MAML2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686N0150/KIAA1819/MAM-3/MAM2/MAM3/MGC176701/MLL-MAML2 | |
MAML2 | |
Recombinant | |
wheat germ expression system |