Learn More
ACROBiosystems Human LTBR / TNFRSF3 Protein, Fc Tag
Marke: ACROBiosystems LTR-H5251-100UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Lymphotoxin-beta receptor (LTBR) is also known as Tumor necrosis factor receptor superfamily member 3 (TNFRSF3), Tumor necrosis factor receptor type III (TNF-RIII), which is a single-pass type I membrane protein containing four TNFR-Cys repeat regions. Except for interacting with HCV core protein, LTBR can not only associate with itself, but also can associate with TRAF3, TRAF4 and TRAF5. As the receptor for the heterotrimeric lymphotoxin containing LTA and LTB, and for TNFS14/LIGHT, LTBR promotes apoptosis via TRAF3 and TRAF5. Furthermore, LTBR may play a role in the development of lymphoid organs.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.4 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002333 | |
100 µg | |
Gln 31 - Met 227 | |
LTBR,D12S370,TNFCR,TNFR3,TNFRSF3,TNFRIII | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.4 kDa | |
Human LTBR, Fc Tag (LTR-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Met 227 (Accession # P36941-1). | |
-20°C | |
LTBR | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |