missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human LTBR / TNFRSF3 Protein, Fc Tag

Marke:  ACROBiosystems LTR-H5251-1MG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827287

  • 1381.76 CHF / 1 Milligramm


Verfügbar ab
28-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Lymphotoxin-beta receptor (LTBR) is also known as Tumor necrosis factor receptor superfamily member 3 (TNFRSF3), Tumor necrosis factor receptor type III (TNF-RIII), which is a single-pass type I membrane protein containing four TNFR-Cys repeat regions. Except for interacting with HCV core protein, LTBR can not only associate with itself, but also can associate with TRAF3, TRAF4 and TRAF5. As the receptor for the heterotrimeric lymphotoxin containing LTA and LTB, and for TNFS14/LIGHT, LTBR promotes apoptosis via TRAF3 and TRAF5. Furthermore, LTBR may play a role in the development of lymphoid organs.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.4 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_002333
HEK293
Human
48.4 kDa
1 mg
Gln 31 - Met 227
LTBR,D12S370,TNFCR,TNFR3,TNFRSF3,TNFRIII
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human LTBR, Fc Tag (LTR-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Met 227 (Accession # P36941-1).
-20°C
LTBR
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation