missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LSP1 Partial ORF (NP_002330.1, 240 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_002330.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 4046 |
Molekulargewicht | 36.63kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16140775
|
Abnova™
H00004046-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 06-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16150775
|
Abnova™
H00004046-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 06-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: TAGRTPKLARQASIELPSMAVASTKSRWETGEVQAQSAAKTPSCKDIVAGDMSKKSLWEQKGGSKTSSTIKSTPSGKRYKFVATGHGKYEKVLVEGGPASpezifikation
NP_002330.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WP34/pp52 | |
LSP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4046 | |
LSP1 (Human) Recombinant Protein (Q01) | |
TAGRTPKLARQASIELPSMAVASTKSRWETGEVQAQSAAKTPSCKDIVAGDMSKKSLWEQKGGSKTSSTIKSTPSGKRYKFVATGHGKYEKVLVEGGPA | |
RUO | |
LSP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |