missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOH11CR2A Full-length ORF (NP_055437.2, 1 a.a. - 786 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00004013-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MVHFCGLLTLHREPVPLKSISVSVNIYEFVAGVSATLNYENEEKVPLEAFFVFPMDEDSAVYSFEALVDGKKIVAELQDKMKARTNYEKAISQGHQAFLLEGDSSSRDVFSCNVGNLQPGSKAAVTLKYVQELPLEADGALRFVLPAVLNPRYQFSGSSKDSCLNVKTPIVPVEDLPYTLSMVATIDSQHGIEKVQSNCPLSPTEYLGEDKTSAQVSLAAGHKFDRDVELLIYYNEVHTPSVVLEMGMPNMKPGHLMGDPSAMVSFYPNIPEDQPSNTCGEFIFLMDRSGSMQSPMSSQDTSQLRIQAAKETLILLLKSLPIGCYFNIYGFGSSYEACFPESVKYTQQTMEEALGRVKLMQADLGGTEILAPLQNIYRGPSIPGHPLQLFVFTDGEVTDTFSVIKEVRINRQKHRCFSFGIGEGTSTSLIKGIARASGGTSEFITGKDRMQSKALRTLKRSLQPVVEDVSLSWHLPPGLSAKMLSPEQTVIFRGQRLISYAQLTGRMPAAETTGEVCLKYTLQGKTFEDKVTFPLQPKPDVNLTIHRLAAKSLLQTKDMGLRETPASDKKDALNLSLESGVISSFTAFIAINKELNKPVQGPLAHRDVPRPILLGASAPLKIKCQSGFRKALHSDRPPSASQPRGELMCYKAKTFQMDDYSLCGLISHKDQHSPGFGENHLVQLIYHQNANGSWDLNEDLAKILGMSLEEIMAAQPAELVDSSGWATILAVIWLHSNGKDLKCEWELLERKAVAWMRAHAGSTMPSVVKAAITFLKSSVDPAIFAFSpezifikation
NP_055437.2 | |
Liquid | |
4013 | |
LOH11CR2A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVHFCGLLTLHREPVPLKSISVSVNIYEFVAGVSATLNYENEEKVPLEAFFVFPMDEDSAVYSFEALVDGKKIVAELQDKMKARTNYEKAISQGHQAFLLEGDSSSRDVFSCNVGNLQPGSKAAVTLKYVQELPLEADGALRFVLPAVLNPRYQFSGSSKDSCLNVKTPIVPVEDLPYTLSMVATIDSQHGIEKVQSNCPLSPTEYLGEDKTSAQVSLAAGHKFDRDVELLIYYNEVHTPSVVLEMGMPNMKPGHLMGDPSAMVSFYPNIPEDQPSNTCGEFIFLMDRSGSMQSPMSSQDTSQLRIQAAKETLILLLKSLPIGCYFNIYGFGSSYEACFPESVKYTQQTMEEALGRVKLMQADLGGTEILAPLQNIYRGPSIPGHPLQLFVFTDGEVTDTFSVIKEVRINRQKHRCFSFGIGEGTSTSLIKGIARASGGTSEFITGKDRMQSKALRTLKRSLQPVVEDVSLSWHLPPGLSAKMLSPEQTVIFRGQRLISYAQLTGRMPAAETTGEVCLKYTLQGKTFEDKVTFPLQPKPDVNLTIHRLAAKSLLQTKDMGLRETPASDKKDALNLSLESGVISSFTAFIAINKELNKPVQGPLAHRDVPRPILLGASAPLKIKCQSGFRKALHSDRPPSASQPRGELMCYKAKTFQMDDYSLCGLISHKDQHSPGFGENHLVQLIYHQNANGSWDLNEDLAKILGMSLEEIMAAQPAELVDSSGWATILAVIWLHSNGKDLKCEWELLERKAVAWMRAHAGSTMPSVVKAAITFLKSSVDPAIFAF | |
RUO | |
VWA5A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
112.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCSC-1/BCSC1/LOH11CR2A | |
VWA5A | |
Yes | |
wheat germ expression system |