missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC91614 Partial ORF (NP_631899, 393 a.a. - 499 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00091614-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: NPSEFKLQKESDNRMVVKRIFSKAIVDNKNLSKGKTDLLVLFLMDHQKDVFKIPGTLHKIVSVKLMAIQNGRDPNRDAGYIYCQRIDQRDYSNNTEKTTKDELLNLLSpezifikation
NP_631899 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NPSEFKLQKESDNRMVVKRIFSKAIVDNKNLSKGKTDLLVLFLMDHQKDVFKIPGTLHKIVSVKLMAIQNGRDPNRDAGYIYCQRIDQRDYSNNTEKTTKDELLNLL | |
RUO | |
DEPDC7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
91614 | |
LOC91614 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TR2/dJ85M6.4 | |
DEPDC7 | |
Recombinant | |
wheat germ expression system |