missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LMP7/PSMB8 ELISA Kit

Artikelnummer. 17814120
Ansicht ändern
Click to view available options
Menge:
10 x 96 Tests
5 x 96 Tests
96 Tests
Packungsgröße:
1 Stück
10 Stück
96 Stück
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
17814120 10 x 96 Tests 10 Stück
17195069 5 x 96 Tests 1 Stück
17114603 96 Tests 96 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17814120 Lieferant Invitrogen™ Lieferanten-Nr. EH303RBX10
  • ONLINE EXCLUSIVE PRICE
    7050.00 CHF
    6838.50 CHF / 10 Stück
    Sparen 211.50 CHF
    3% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 07-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

Human LMP7/PSMB8 quantitates human LMP7/PSMB8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LMP7/PSMB8.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC (major histocompatibility complex). Expression of this gene is induced by gamma interferon and this gene product replaces catalytic subunit 3 (proteasome beta 5 subunit) in the immunoproteasome. Proteolytic processing is required to generate a mature subunit. Two alternative transcripts encoding two isoforms have been identified; both isoforms are processed to yield the same mature subunit.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; Große multifunktionale Protease 7; LMP7; LMP-7; niedermolekulares Polypeptid 7; niedermolekulares Massenprotein 7; Protein mit niedrigem Molekulargewicht 7; Makropain-Untereinheit C13; Mc13; MC13 (LMP7); MGC1491; Multikatalytische Endopeptidase-Komplex-Untereinheit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; Protease-Komponente C13; Proteasom-Untereinheit (Prosome, Makropin), Beta-Typ 8 (große multifunktionale Peptidase 7); Proteasom-Untereinheit (Prosome, Makropin), Beta-Typ, 8; Proteasom-Untereinheit (Prosome, Makropin), Beta-Typ, 8 (große multifunktionale Peptidase 7); Proteasom-katalytische Untereinheit 3 i; Proteasom-Komponente C13; Proteasom-Untereinheit beta 5 i; Proteasom-Untereinheit beta 8; Proteasom-Untereinheit beta Typ 8; Proteasom-Untereinheit beta-8; Proteasom-Untereinheit beta-5 i; Proteasom-Untereinheit MC13; Proteasom-Einheit Y2; Proteasom-Untereinheit; Proteasom-Einheit; Proteasomeinheit Y2; Proteasom-Untereinheit; Proteasomeinheit; Proteosom (Prosome, Makropain) Untereinheit, Beta-Typ 8 (große multifunktionale Peptidase 7); Proteosome Beta 8-Untereinheit; PSMB5i; PSMB8; wirklich interessantes neues Gen 10-Protein; RING10; Y2
Assay-Bereich 2,2 bis 500 ng/ml
Assay-Empfindlichkeit: 2,2 ng/ml
Konjugat Biotin
Immunassay-Kit-Format Sandwich ELISA Kit
Produkttyp ELISA
Probentyp Plasma, Serum, Überstand
Zielspezies Human
Zur Verwendung mit (Geräte) Kolorimetrisches Mikrotiterplatten-Lesegerät
Gen-Alias ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; large multifunctional protease 7; Lmp7; Lmp-7; low molecular mass polypeptide 7; low molecular mass protein 7; low molecular weight protein 7; macropain subunit C13; Mc13; MC13 (LMP7); MGC1491; multicatalytic endopeptidase complex subunit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; protease component C13; proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteasome (prosome, macropain) subunit, beta type, 8; proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7); proteasome catalytic subunit 3i; proteasome component C13; proteasome subunit beta 5i; proteasome subunit beta 8; proteasome subunit beta type 8; proteasome subunit beta type-8; Proteasome subunit beta-5i; proteasome subunit MC13; proteasome subunit Y2; proteasome-related gene 7; proteosome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteosome beta 8 subunit; PSMB5i; Psmb8; Really interesting new gene 10 protein; Ring10; Y2ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; large multifunctional protease 7; Lmp7; Lmp-7; low molecular mass polypeptide 7; low molecular mass protein 7; low molecular weight protein 7; macropain subunit C13; Mc13; MC13 (LMP7); MGC1491; multicatalytic endopeptidase complex subunit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; protease component C13; proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteasome (prosome, macropain) subunit, beta type, 8; proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7); proteasome catalytic subunit 3i; proteasome component C13; proteasome subunit beta 5i; proteasome subunit beta 8; proteasome subunit beta type 8; proteasome subunit beta type-8; Proteasome subunit beta-5i; proteasome subunit MC13; proteasome subunit Y2; proteasome-related gene 7; proteosome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteosome beta 8 subunit; PSMB5i; Psmb8; Really interesting new gene 10 protein; Ring10; Y2;
Gen-ID (Entrez) P28062
Gensymbol Psmb8
Interassay-VK < 12%
Intraassay-VK < 10%
Kitinhalt Vorbeschichtete 96-Well-Platte, Standard, Assay-Verdünnerkonzentrat, Biotinylierter Nachweisantikörper, SAV-HRP, Waschpuffer, Chromogen, Stopplösung, Dichtungsfolien für Mikrotiterplatten, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Marker oder Farbstoff HRP
Menge 10 x 96 Tests
Kennzeichnung Nur für Forschungszwecke
Probenvolumen Überstand, 50 μl; Serum, 50 μl; Plasma, 50 μl
Versandbedingung Wet or Dry Ice
Lagerungsbedingungen 2 bis 8 °C
Ziel PSMB8
Prüfzeit 1 h 20 min
Gesamte Assay-Zeit 4 h 45 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.