missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LIMS1 Partial ORF (NP_004978.2, 226 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

361.00 CHF - 547.00 CHF

Spezifikation

Zugriffsnummer NP_004978.2
Zur Verwendung mit (Anwendung) Antibody Production, ELISA, Protein Array, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 3987
Molekulargewicht 36.74kDa
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16140705
Unterlagen ansehen Aktionsdetails
Abnova™
H00003987-Q01.25UG
25 ug
547.00 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16130705
Unterlagen ansehen Aktionsdetails
Abnova™
H00003987-Q01.10UG
10 ug
361.00 CHF
10 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The protein encoded by this gene is an adaptor protein which contains five LIM domains, or double zinc fingers. The protein is likely involved in integrin signaling through its LIM domain-mediated interaction with integrin-linked kinase, found in focal adhesion plaques. It is also thought to act as a bridge linking integrin-linked kinase to NCK adaptor protein 2, which is involved in growth factor receptor kinase signaling pathways. Its localization to the periphery of spreading cells also suggests that this protein may play a role in integrin-mediated cell adhesion or spreading. [provided by RefSeq]

Sequence: FLGHRHYERKGLAYCETHYNQLFGDVCFHCNRVIEGDVVSALNKAWCVNCFACSTCNTKLTLKNKFVEFDMKPVCKKCYEKFPLELKKRLKKLAETLGRK
Spezifikation

Spezifikation

NP_004978.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PINCH/PINCH1
LIMS1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
3987
LIMS1 (Human) Recombinant Protein (Q01)
FLGHRHYERKGLAYCETHYNQLFGDVCFHCNRVIEGDVVSALNKAWCVNCFACSTCNTKLTLKNKFVEFDMKPVCKKCYEKFPLELKKRLKKLAETLGRK
RUO
LIMS1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications