Learn More
Abnova™ Human LIMS1 Full-length ORF (NP_004978.2, 1 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
The protein encoded by this gene is an adaptor protein which contains five LIM domains, or double zinc fingers. The protein is likely involved in integrin signaling through its LIM domain-mediated interaction with integrin-linked kinase, found in focal adhesion plaques. It is also thought to act as a bridge linking integrin-linked kinase to NCK adaptor protein 2, which is involved in growth factor receptor kinase signaling pathways. Its localization to the periphery of spreading cells also suggests that this protein may play a role in integrin-mediated cell adhesion or spreading. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_004978.2 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 3987 |
Molekulargewicht | 63.7kDa |
Name | LIMS1 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 ug |
Immunogen | MANALASATCERCKGGFAPAEKIVNSNGELYHEQCFVCAQCFQQFPEGLFYEFEGRKYCEHDFQMLFAPCCHQCGEFIIGRVIKAMNNSWHPECFRCDLCQEVLADIGFVKNAGRHLCRPCHNREKARGLGKYICQKCHAIIDEQPLIFKNDPYHPDHFNCANCGKELTADARELKGELYCLPCHDKMGVPICGACRRPIEGRVVNAMGKQWHVEHFVCAKCEKPFLGHRHYERKGLAYCETHYNQLFGDVCFHCNRVIEGDVVSALNKAWCVNCFACSTCNTKLTLKNKFVEFDMKPVCKKCYEKFPLELKKRLKKLAETLGRK |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.