missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LGP1 Partial ORF (NP_115873, 431 a.a. - 528 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084514-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: SSAGSAPHYEVFVALRGLRNLSEENRDKLDHCLQEASPRYKSLRFWGSVGPARVHLVGQGAFRALRAALAACPSSPFPPAMPRVLRHRHLAQCLQERVSpezifikation
NP_115873 | |
Liquid | |
84514 | |
LGP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D11LGP1/LGP1 | |
GHDC | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SSAGSAPHYEVFVALRGLRNLSEENRDKLDHCLQEASPRYKSLRFWGSVGPARVHLVGQGAFRALRAALAACPSSPFPPAMPRVLRHRHLAQCLQERV | |
RUO | |
GHDC | |
Wheat Germ (in vitro) | |
GST |