Learn More
Abnova™ Human LAMA2 Partial ORF (NP_000417, 3013 a.a. - 3122 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
Laminin, an extracellular protein, is a major component of the basement membrane. It is thought to mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. It is composed of three subunits, alpha, beta, and gamma, which are bound to each other by disulfide bonds into a cross-shaped molecule. This gene encodes the alpha 2 chain, which constitutes one of the subunits of laminin 2 (merosin) and laminin 4 (s-merosin). Mutations in this gene have been identified as the cause of congenital merosin-deficient muscular dystrophy. Two transcript variants encoding different proteins have been found for this gene. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_000417 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 3908 |
Molekulargewicht | 37.84kDa |
Name | LAMA2 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 ug |
Immunogen | DAGVPGHLCDGQWHKVTANKIKHRIELTVDGNQVEAQSPNPASTSADTNDPVFVGGFPDDLKQFGLTTSIPFRGCIRSLKLTKGTGKPLEVNFAKALELRGVQPVSCPAN |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Produktvorschläge
Customers who viewed this item also viewed.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.