missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA0746 Full-length ORF (NP_056002.1, 1 a.a. - 979 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00023231-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16127082

  • 379.00 CHF / 10 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MVYRDDYFIRHSISVSAVIVRAWITHKYSGRDWNVKWEENLLHAVAKNYTLLQTIPPFERPFKDHQVCLEWNMGYIWNLRANRIPQCPLENDVVALLGFPYASSGENTGIVKKFPRFRNRELEATRRQRMDYPVFTVSLWLYLLHYCKANLCGILYFVDSNEMYGTPSVFLTEEGYLHIQMHLVKGEDLAVKTKFIIPLKEWFRLDISFNGGQIVVTTSIGQDLKSYHNQTISFREDFHYNDTAGYFIIGGSRYVAGIEGFFGPLKYYRLRSLHPAQIFNPLLEKQLAEQIKLYYERCAEVQEIVSVYASAAKHGGERQEACHLHNSYLDLQRRYGRPSMCRAFPWEKELKDKHPSLFQALLEMDLLTVPRNQNESVSEIGGKIFEKAVKRLSSIDGLHQISSIVPFLTDSSCCGYHKASYYLAVFYETGLNVPRDQLQGMLYSLVGGQGSERLSSMNLGYKHYQGIDNYPLDWELSYAYYSNIATKTPLDQHTLQGDQAYVETIRLKDDEILKVQTKEDGDVFMWLKHEATRGNAAAQQRLAQMLFWGQQGVAKNPEAAIEWYAKGALETEDPALIYDYAIVLFKGQGVKKNRRLALELMKKAASKGLHQAVNGLGWYYHKFKKNYAKAAKYWLKAEEMGNPDASYNLGVLHLDGIFPGVPGRNQTLAGEIFHKAAQGGHMEGTLWCSLYYITGNLETFPRDPEKAVVWAKHVAEKNGYLGHVIRKGLNAYLEGSWHEALLYYVLAAETGIEVSQTNLAHICEERPDLARRYLGVNCVWRYYNFSVFQIDAPSFAYLKMGDLYYYGHQNQSQDLELSVQMYAQAALDGDSQGFFNLALLIEEGTIIPHHILDFLEIDSTLHSNNISILQELYERCWSHSNEESFSPCSLAWLYLHLRLLWGAILHSALIYFLGTFLLSILIAWTVQYFQSVSASDPPPRPSQASPDTATSTASPAVTPAADASDQDQPTVTNNPEPRG
Spezifikation

Spezifikation

NP_056002.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
138.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781J1697/FLJ21629/FLJ41299
KIAA0746
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23231
KIAA0746 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVYRDDYFIRHSISVSAVIVRAWITHKYSGRDWNVKWEENLLHAVAKNYTLLQTIPPFERPFKDHQVCLEWNMGYIWNLRANRIPQCPLENDVVALLGFPYASSGENTGIVKKFPRFRNRELEATRRQRMDYPVFTVSLWLYLLHYCKANLCGILYFVDSNEMYGTPSVFLTEEGYLHIQMHLVKGEDLAVKTKFIIPLKEWFRLDISFNGGQIVVTTSIGQDLKSYHNQTISFREDFHYNDTAGYFIIGGSRYVAGIEGFFGPLKYYRLRSLHPAQIFNPLLEKQLAEQIKLYYERCAEVQEIVSVYASAAKHGGERQEACHLHNSYLDLQRRYGRPSMCRAFPWEKELKDKHPSLFQALLEMDLLTVPRNQNESVSEIGGKIFEKAVKRLSSIDGLHQISSIVPFLTDSSCCGYHKASYYLAVFYETGLNVPRDQLQGMLYSLVGGQGSERLSSMNLGYKHYQGIDNYPLDWELSYAYYSNIATKTPLDQHTLQGDQAYVETIRLKDDEILKVQTKEDGDVFMWLKHEATRGNAAAQQRLAQMLFWGQQGVAKNPEAAIEWYAKGALETEDPALIYDYAIVLFKGQGVKKNRRLALELMKKAASKGLHQAVNGLGWYYHKFKKNYAKAAKYWLKAEEMGNPDASYNLGVLHLDGIFPGVPGRNQTLAGEIFHKAAQGGHMEGTLWCSLYYITGNLETFPRDPEKAVVWAKHVAEKNGYLGHVIRKGLNAYLEGSWHEALLYYVLAAETGIEVSQTNLAHICEERPDLARRYLGVNCVWRYYNFSVFQIDAPSFAYLKMGDLYYYGHQNQSQDLELSVQMYAQAALDGDSQGFFNLALLIEEGTIIPHHILDFLEIDSTLHSNNISILQELYERCWSHSNEESFSPCSLAWLYLHLRLLWGAILHSALIYFLGTFLLSILIAWTVQYFQSVSASDPPPRPSQASPDTATSTASPAVTPAADASDQDQPTVTNNPEPRG
RUO
KIAA0746
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote