missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KCTD18 Full-length ORF (NP_689600.2, 1 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00130535-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16127893

  • 364.00 CHF / 10 Mikrogramm

Verfügbar ab: 03-05-2023
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MEGHKAEEEVLDVLRLNVGGCIYTARRESLCRFKDSMLASMFSGRFPLKTDESGACVIDRDGRLFKYLLDYLHGEVQIPTDEQTRIALQEEADYFGIPYPYSLSDHLANEMETYSLRSNIELKKALTDFCDSYGLVCNKPTVWVLHYLNTSGASCESRIIGVYATKTDGTDAIEKQLGGRIHSKGIFKREAGNNVQYIWSYYSVAELKKMMDAFDAWEGKGVSYWRVPHELIECWTLEERPLLGSLRHMAPIRKRRLITFNEADESVNYKTGPKPVRFLGPSTSTQIKVKNSASVTVSPASAIQTSAGATANRFQSGSRRKAAQRSAPSRATALVGTGAPGHPQASPGAASAENGGTHLPPAKVLLSDKKPTPQRVIKLKRTPLCATAPCLPSPTATRQANSLKPLPGEAARALGVRTENGKNKGN
Spezifikation

Spezifikation

NP_689600.2
Liquid
130535
KCTD18 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGHKAEEEVLDVLRLNVGGCIYTARRESLCRFKDSMLASMFSGRFPLKTDESGACVIDRDGRLFKYLLDYLHGEVQIPTDEQTRIALQEEADYFGIPYPYSLSDHLANEMETYSLRSNIELKKALTDFCDSYGLVCNKPTVWVLHYLNTSGASCESRIIGVYATKTDGTDAIEKQLGGRIHSKGIFKREAGNNVQYIWSYYSVAELKKMMDAFDAWEGKGVSYWRVPHELIECWTLEERPLLGSLRHMAPIRKRRLITFNEADESVNYKTGPKPVRFLGPSTSTQIKVKNSASVTVSPASAIQTSAGATANRFQSGSRRKAAQRSAPSRATALVGTGAPGHPQASPGAASAENGGTHLPPAKVLLSDKKPTPQRVIKLKRTPLCATAPCLPSPTATRQANSLKPLPGEAARALGVRTENGKNKGN
RUO
KCTD18
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
6530404F10Rik/FLJ31322/FLJ37818
KCTD18
Yes
wheat germ expression system
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote