missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KCNJ15 Partial ORF (NP_002234, 290 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00003772-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16150395

  • 458.00 CHF / 25 Mikrogramm


Verfügbar ab
11-08-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: TSAVCQSRTSYIPEEIYWGFEFVPVVSLSKNGKYVADFSQFEQIRKSPDCTFYCADSEKQQLEEKY
Spezifikation

Spezifikation

NP_002234
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TSAVCQSRTSYIPEEIYWGFEFVPVVSLSKNGKYVADFSQFEQIRKSPDCTFYCADSEKQQLEEKY
RUO
KCNJ15
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3772
KCNJ15 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IRKK|KIR1.3|KIR4.2|MGC13584
KCNJ15
Yes
Videos
Dokumentation