Learn More
Abnova™ Human KCNH7 Partial ORF (NP_775185.1, 132 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00090134-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. There are at least two alternatively spliced transcript variants derived from this gene and encoding distinct isoforms. [provided by RefSeq]
Sequence: VTDNENAATPERVNPILPIKTVNRKFFGFKFPGLRVLTYRKQSLPQEDPDVVVIDSSKHSDDSVAMKHFKSPTKESCSPSEADDTKALIQPSKCSPLVNSpezifikation
NP_775185.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VTDNENAATPERVNPILPIKTVNRKFFGFKFPGLRVLTYRKQSLPQEDPDVVVIDSSKHSDDSVAMKHFKSPTKESCSPSEADDTKALIQPSKCSPLVN | |
RUO | |
KCNH7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
90134 | |
KCNH7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERG3/HERG3/Kv11.3/MGC45986 | |
KCNH7 | |
Recombinant | |
wheat germ expression system |