Learn More
Abnova™ Human KCNE2 Full-length ORF (NP_751951.1, 1 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009992-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, isk-related subfamily. This member is a small integral membrane subunit that assembles with the KCNH2 gene product, a pore-forming protein, to alter its function. This gene is expressed in heart and muscle and the gene mutations are associated with cardiac arrhythmia. [provided by RefSeq]
Sequence: MSTLSNFTQTLEDVFRRIFITYMDNWRQNTTAEQEALQAKVDAENFYYVILYLMVMIGMFSFIIVAILVSTVKSKRREHSNDPYHQYIVEDWQEKYKSQILNLEESKATIHENIGAAGFKMSPSpezifikation
NP_751951.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LQT5/LQT6/MGC138292/MIRP1 | |
KCNE2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9992 | |
KCNE2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTLSNFTQTLEDVFRRIFITYMDNWRQNTTAEQEALQAKVDAENFYYVILYLMVMIGMFSFIIVAILVSTVKSKRREHSNDPYHQYIVEDWQEKYKSQILNLEESKATIHENIGAAGFKMSP | |
RUO | |
KCNE2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |