missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Jagged 1 / JAG1 Protein, His Tag
365.69 CHF - 3076.85 CHF
Spezifikation
Zugriffsnummer | NP_000205.1 |
---|---|
Molekulargewicht | 112.3 kDa |
Quelle | Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Ser 1046 (Accession # P78504-1). |
Immunogen | Gln 34 - Ser 1046 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15837167
|
ACROBiosystems
JA1-H52H9-50UG |
50 μg |
365.69 CHF
50 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827167
|
ACROBiosystems
JA1-H52H9-1MG |
1 mg |
3076.85 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Protein jagged-1 (JAG1) is also known as Jagged1, hJ1, JAGL1 and CD339, which is a single-pass type I membrane protein containing one DSL domain and 15 EGF-like domains. JAG1 is widely expressed in adult and fetal tissues,the expression of JAG1 is up-regulated in cervical squamous cell carcinoma and also expressed in bone marrow cell line HS-27a.JAG1 is a ligand for multiple Notch receptors and involved in the mediation of Notch signaling. JAG1 is involved in early and late stages of mammalian cardiovascular development. JAG1 inhibits myoblast differentiation by similarity and enhances fibroblast growth factor-induced angiogenesis (in vitro).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 112.3 kDa. The protein migrates as 130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000205.1 | |
Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Ser 1046 (Accession # P78504-1). | |
-20°C | |
Jagged 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
112.3 kDa | |
Gln 34 - Ser 1046 | |
CD339,JAG1,Jagged1,hJ1,JAGL1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |