Learn More
ACROBiosystems Human Integrin alpha V beta 8 (ITGAV&ITGB8) Heterodimer Protein, His Tag&Tag Free
Marke:  ACROBiosystems IT8-H52W4-1MG
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15817167
  • 3316.22 CHF / 1 Milligramm
Verfügbar ab
04-06-2021
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Integrin alpha V beta 8 (ITGAV & ITGB8 or ITGAVB8) is expressed in yolk sac, placenta, brain perivascular astrocytes, Schwann cells, renal glomerular mesangial cells and pulmonary epithelial cells. Unlike other alpha V integrins, ITGAVB8 does not appear to assume different activation states, and the cytoplasmic tail does not connect to the cytoskeleton. It binds ligands containing an RGD motif, including vitronectin, fibrin and the latency associated peptide (LAP) of the latent TGF-beta complex. High affinity binding of alpha V beta 8 to LAP allows proteolytic cleavage by MT1-MMP, which releases active TGF-beta. This mechanism differs from that of alpha V beta 6, the other alpha V integrin which can activate TGF-beta from latency through non-proteolytic mechanisms. Downstream effects of TGF-beta activation include control of cell growth and associated vascularization.
Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB8, has a calculated MW of 112.9 kDa (ITGAV) and 76.5 kDa (ITGB8). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidic
Spezifikation
Spezifikation
NP_002201.1 (ITGAV) and NP_002205.1 (ITGB8)
HEK293
Human
112.9 kDa (ITGAV) and 76.5 kDa (ITGB8)
1 mg
Phe 31 - Val 992 (ITGAV) and Glu 43 - Arg 684 (ITGB8)
Integrin alpha V beta 8,ITGAVandITGB8
Unconjugated
50 mM Tris, 100 mM NaCl, pH8.0
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human ITGAV&ITGB8; Heterodimer Protein, His Tag&Tag; Free (IT8-H52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) and Glu 43 - Arg 684 (ITGB8) (Accession # NP_002201.1 (ITGAV) and NP_002205.1 (ITGB8)).
-20°C
Integrin alpha V beta 8
His Tag