Learn More
ACROBiosystems Human Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein, His Tag&Tag Free
517.01 CHF - 4035.20 CHF
Spezifikation
Zugriffsnummer | NP_002201.1 (ITGAV) and NP_002204.2 (ITGB5) |
---|---|
Molekulargewicht | 112.9 kDa (ITGAV) and 81.9 kDa (ITGB5) |
Quelle | Human ITGAV&ITGB5; Heterodimer Protein, His Tag&Tag; Free (IT5-H52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) and Gly 24 - Asn 719 (ITGB5) (Accession # NP_002201.1 (ITGAV) and NP_002204.2 (ITGB5)). |
Immunogen | Phe 31 - Val 992 (ITGAV) and Gly 24 - Asn 719 (ITGB5) |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827157
|
ACROBiosystems
IT5-H52W5-1MG |
1 mg |
4035.20 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817157
|
ACROBiosystems
IT5-H52W5-100UG |
100 μg |
517.01 CHF
100 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Integrin alpha V beta 5 (ITGAV & ITGB5) is expressed on a wide variety of cell types including keratinocytes, fibroblasts, adhesive monocytes, embryonic stem cells, and select endothelium and epithelium. ITGAV & ITGB5 binds ligands containing an RGD motif, notably vitronectin. Growth factors that increase PKC activity, such as VEGF or TGF alpha, promote ITGAV & ITGB5-mediated angiogenesis while alpha V beta 3, which may be expressed in the same cell, responds to FGF-basic and TNF alpha. An inhibitor of both down regulates tumor angiogenesis. During lung inflammation, up regulation of ITGAV & ITGB5 on myofibroblasts or infiltrating lymphocytes may contribute to fibrosis by freeing TGF beta from latency.
Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB5, has a calculated MW of 112.9 kDa (ITGAV) and 81.9 kDa (ITGB5). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidicSpezifikation
NP_002201.1 (ITGAV) and NP_002204.2 (ITGB5) | |
Human ITGAV&ITGB5; Heterodimer Protein, His Tag&Tag; Free (IT5-H52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) and Gly 24 - Asn 719 (ITGB5) (Accession # NP_002201.1 (ITGAV) and NP_002204.2 (ITGB5)). | |
-20°C | |
Integrin alpha V beta 5 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
112.9 kDa (ITGAV) and 81.9 kDa (ITGB5) | |
Phe 31 - Val 992 (ITGAV) and Gly 24 - Asn 719 (ITGB5) | |
Integrin alpha V beta 5,ITGAVandITGB5 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |