missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha V beta 3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
262.00 CHF - 526.00 CHF
Spezifikation
| Antigen | Integrin alpha V beta 3 |
|---|---|
| Klon | 2549B |
| Verdünnung | Flow Cytometry 0.25 ug/10^6 cells |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18774017
![]() |
R&D Systems
MAB12192-SP |
25 μg |
262.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18752397
![]() |
R&D Systems
MAB12192-100 |
100 μg |
526.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Spezifikation
| Integrin alpha V beta 3 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
| Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 3, Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 3 (Gly27-Asp718), Accession # NP_002201 and AAA52589 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2549B | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| NP_002201 and AAA52589 | |
| 3685 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts