missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein, His Tag&Tag Free
517.01 CHF - 4035.20 CHF
Spezifikation
Zugriffsnummer | NP_000201.2 (ITGA6) and NP_002202.2 (ITGB1) |
---|---|
Molekulargewicht | 116.8 kDa (ITGA6) and 83.7 kDa (ITGB1) |
Quelle | Human ITGA6&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 24 - Gly 1012 (ITGA6) and Gln 21 - Asp 728 (ITGB1) (Accession # P23229-2 (ITGA6) and P05556-1 (ITGB1)). |
Immunogen | Phe 24 - Gly 1012 (ITGA6) and Gln 21 - Asp 728 (ITGB1) |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827147
|
ACROBiosystems
IT1-H52W7-1MG |
1 mg |
4035.20 CHF
1 Milligramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817147
|
ACROBiosystems
IT1-H52W7-100UG |
100 μg |
517.01 CHF
100 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Integrin alpha 6 beta 1, also called platelet glycoprotein GPIc-IIa, is a receptor for laminin on platelets. The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. ITGA6 has been shown to interact with TSPAN4 and GIPC1.
Human ITGA6&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA6 and ITGB1, has a calculated MW of 116.8 kDa (ITGA6) and 83.7 kDa (ITGB1). Subunit ITGA6 is fused with an acidic tail at the C-terminus and followed by a polyhistidSpezifikation
NP_000201.2 (ITGA6) and NP_002202.2 (ITGB1) | |
Human ITGA6&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 24 - Gly 1012 (ITGA6) and Gln 21 - Asp 728 (ITGB1) (Accession # P23229-2 (ITGA6) and P05556-1 (ITGB1)). | |
-20°C | |
Integrin alpha 6 beta 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
116.8 kDa (ITGA6) and 83.7 kDa (ITGB1) | |
Phe 24 - Gly 1012 (ITGA6) and Gln 21 - Asp 728 (ITGB1) | |
Integrin alpha 6 beta 1,ITGA6andITGB1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |