Learn More
ACROBiosystems Human IL-7 R alpha / CD127 Protein, Mouse IgG2a Fc Tag
Marke: ACROBiosystems IL7-H5258-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Interleukin-7 receptor subunit alpha (IL7Ra) is also known as CD antigen CD127, which belongs to the type I cytokine receptor family and type 4 subfamily. IL7Ra /CD127 contains 1 fibronectin type-III domain. IL7Ra /CD127 is expressed on various cell types, including naive and memory T cells and many others. IL7Ra /CD127 is receptor for interleukin-7 and also acts as a receptor for thymic stromal lymphopoietin (TSLP). The IL7 receptor is a heterodimer of IL7R and IL2RG. The TSLP receptor is a heterodimer of CRLF2 and IL7R.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 52.0 kDa. The protein migrates as 65-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
52.0 kDa | |
Human IL-7 R alpha, Mouse IgG2a Fc Tag (IL7-H5258) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Gly 236 (Accession # P16871-1). | |
-20°C | |
IL-7 R alpha | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
1 mg | |
Glu 21 - Gly 236 | |
IL7Ra,CD127 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |