Learn More
ACROBiosystems Human IL-27 Ra / TCCR Protein, Fc Tag
Marke:  ACROBiosystems ILA-H5254-100UG
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15827077
  • 312.65 CHF / 100 Mikrogramm
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Interleukin-27 receptor subunit alpha ( IL27RA) is also known as Cytokine receptor WSX-1, Cytokine receptor-like 1, Type I T-cell cytokine receptor (TCCR), ZcytoR1, IL27RA belongs to the type I cytokine receptor family and type 2 subfamily. IL27RA contains three fibronectin type-III domains. IL27RA is highly expressed in lymphoid tissues such as spleen, lymph nodes and peripheral blood leukocytes. IL27RA is receptor for IL27, but requires IL6ST/gp130 to mediate signal transduction in response to IL27. IL27RA involved in the regulation of Th1-type immune responses. IL27RA also appears to be involved in innate defense mechanisms.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 78.9 kDa. The protein migrates as 100-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH28003
100 μg
Gln 33 - Arg 514
IL27RA,CRL1,TCCR,WSX1
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
78.9 kDa
Human IL-27 Ra, Fc Tag (ILA-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Arg 514 (Accession # AAH28003).
-20°C
IL-27 Ra
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens