missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-23 Recombinant Protein, PeproTech®

Artikelnummer. p-7246355
Click to view available options
Menge:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
2 Mikrogramm
2 Stück
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17890433

Marke: Gibco™ 20023100UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

200-23-1MG will be provided as 2 x 500 μg (200-23-500UG). Recombinant Human IL-23 is a 53.5 kDa, heterodimeric protein consisting of the two subunits, p19 (170 amino acids) and p40 (306 amino acids). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 that is disulfide-linked with to two subunits, a p19 subunit unique to IL-23, and a p40 subunit that is shared with IL-12. IL-23 interacts with IL-12Rbeta1 and a beta2-like receptor subunit with STAT4 binding domain, IL-23R. IL-23 is secreted by activated mouse and human dendritic cells and is involved in the inflammation process and associated with autoimmune diseases. Mouse IL-23 was found not to induce significant amounts of IFN-gamma, induces strong proliferation of memory T cells (but not naive T cells), and activate mouse memory T cells to produce the proinflammatory cytokine IL-17. On the other hand, human IL-23 induces proliferation of memory T cells, induces moderate levels of IFN-gamma, and is produced by naive and memory T cells. IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance, and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines. IL-23 interacts with the receptors IL-12R beta1 and IL-23R to activate the Jak-Stat signaling cascade. IL-23 has been shown to possess potent anti-tumor and anti-metastatic activity in mouse models of cancer, suggesting a potential role for IL-23 in therapeutic treatment of cancer.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q9NPF7
Zur Verwendung mit (Anwendung) Functional Assay
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 51561
Molekulargewicht 53.5 kDa
Name Human IL-23
Menge 100 μg
Quelle Insect cells
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/ μg
Gen-Alias IL23; IL-23; IL-23 subunit alpha; IL23A; IL-23 A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; P19; SGRF; UNQ2498/PRO5798
Gebräuchliche Bezeichnung IL-23
Gensymbol IL23A
Biologische Aktivität Measured by its ability to induce IL-17 secretion by mouse splenocytes.
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz p19 subunit: RAVPGGSSPA WTQCQQLSQK LCTLAWSAHP LVGHMDLREE GDEETTNDVP HIQCGDGCDP QGLRDNSQFC LQRIHQGLIF YEKLLGSDIF TGEPSLLPDS PVGQLHASLL GLSQLLQPEG HHWETQQIPS LSPSQPWQRL LLRFKILRSL QAFVAVAARV FAHGAATLSP p40 subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
Inhalt und Lagerung -20°C
Expressionssystem Insect cells
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt