missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-22 R alpha 1 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Marke: R&D Systems FAB2770P
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-22R alpha 1 Monoclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for Flow Cytometry.
Spezifikation
| IL-22R alpha 1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58985 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 305405 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q8N6P7 | |
| IL22RA1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 Accession # Q8N6P7 | |
| 100 Tests | |
| Primary | |
| Detects human IL-22 R alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-10, rhIL-22BP or rhIL-20 R alpha is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur