missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human IL-18 R1 / CD218a Protein, Fc Tag
365.69 CHF - 1374.49 CHF
Spezifikation
Zugriffsnummer | AAH69575 |
---|---|
Molekulargewicht | 62.4 kDa |
Quelle | Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Arg 329 (Accession # AAH69575). |
Immunogen | Ala 19 - Arg 329 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807027
|
ACROBiosystems
IL1-H5259-1MG |
1 mg |
1374.49 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817027
|
ACROBiosystems
IL1-H5259-200UG |
200 μg |
365.69 CHF
200 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Interleukin-18 receptor 1 (IL18R1) is also known as CD218 antigen-like family member A (CD218a), IL1 receptor-related protein (IL-1Rrp or IL1R-rp), CDw218a, IL18Ra, IL1RRP, which belongs to the interleukin-1 receptor family. IL18R1 contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL18R1 is receptor for interleukin 18 (IL-18). IL18R1 binds to the agonist leads to the activation of NF-kappa-B, but does not bind IL1A/interleukin-1 alpha or IL1B/interleukin-1 beta.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH69575 | |
Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Arg 329 (Accession # AAH69575). | |
-20°C | |
IL-18 R1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
62.4 kDa | |
Ala 19 - Arg 329 | |
IL18R1,IL1RRP,CD218a,IL18Ra | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |