Learn More
ACROBiosystems Human IL-17E / IL-25 Protein, His Tag
Marke: ACROBiosystems IL5-H4221-20UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Interleukin 25 (IL25) is also known as Interleukin-17E (IL-17E), is a distinct member of the IL17 cytokine family comprised of at least six members sharing a conserved cysteine-knot structure but divergent at the N-terminus. IL-25 is secreted by type 2 helper T cells (Th2) and mast cells. IL25 can induce NF-κB activation, and stimulate the production of IL8. Both this cytokine and IL17B are ligands for the cytokine receptor IL17RB. IL-25 induces the production of other cytokines, including IL-4, IL-5 and IL-13 in multiple tissues, which stimulate the expansion of eosinophils. This cytokine is an important molecule controlling immunity of the gut and has been implicated in chronic inflammation associated with the gastrointestinal tract. IL-25 can kill some types of breast cancer cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.6 kDa. The protein migrates as 23-28 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
NP_073626.1 | |
20 μg | |
Tyr 33 - Gly 177 | |
IL-17 E,IL-25,Interleukin-25 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
17.6 kDa | |
Human IL-17 E, His Tag (IL5-H4221) is expressed from human 293 cells (HEK293). It contains AA Tyr 33 - Gly 177 (Accession # NP_073626.1). | |
-20°C | |
IL-17 E | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |