Learn More
ACROBiosystems Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag
214.38 CHF - 5737.57 CHF
Spezifikation
Zugriffsnummer | NP_002178.2 (IL12B) and NP_000873.2 (IL12A) |
---|---|
Molekulargewicht | 36.6 kDa (IL12B) and 23.8 kDa (IL12A) |
Quelle | Human IL-12 B&IL-12; A Heterodimer Protein, His Tag&Flag; Tag (IL2-H4210) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (IL-12 B) and Arg 23 - Ser 219 (IL-12 A) (Accession # P29460-1 (IL12B) and P29459-1 (IL12A)). |
Immunogen | Ile 23 - Ser 328 (IL-12 B) and Arg 23 - Ser 219 (IL-12 A) |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817037
|
ACROBiosystems
IL2-H4210-50UG |
50 μg |
479.18 CHF
50 Mikrogramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15897027
|
ACROBiosystems
IL2-H4210-1MG |
1 mg |
5737.57 CHF
1 Milligramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15807037
|
ACROBiosystems
IL2-H4210-20UG |
20 μg |
214.38 CHF
20 Mikrogramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Interleukin 12 (IL12) is also known as p70, and is an interleukin that is naturally produced by dendritic cells, macrophages and human B-lymphoblastoid cells (NC-37) in response to antigenic stimulation.IL12 is a heterodimeric cytokine, containing IL-12A (p35) and IL-12B (p40). IL-12 is involved in the differentiation of naive T cells into Th1 cells. It is known as a T cell-stimulating factor, which can stimulate the growth and function of T cells. It stimulates the production of IFN-γ and TNF-α from T cells and NK cells, and reduces IL-4 mediated suppression of IFN-γ. IL-12 plays an important role in the activities of natural killer cells and T lymphocytes. IL-12 also has anti-angiogenic activity, which means it can block the formation of new blood vessels.
Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of IL-12B and IL-12A, has a calculated MW of 36.6 kDa (IL-12B) and 23.8 kDa (IL-12A). Subunit IL-12B is fused with polyhistidine tag at the C-terminus and subunit IL-12Spezifikation
NP_002178.2 (IL12B) and NP_000873.2 (IL12A) | |
Human IL-12 B&IL-12; A Heterodimer Protein, His Tag&Flag; Tag (IL2-H4210) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (IL-12 B) and Arg 23 - Ser 219 (IL-12 A) (Accession # P29460-1 (IL12B) and P29459-1 (IL12A)). | |
-20°C | |
IL-12 B and IL-12 A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
36.6 kDa (IL12B) and 23.8 kDa (IL12A) | |
Ile 23 - Ser 328 (IL-12 B) and Arg 23 - Ser 219 (IL-12 A) | |
IL12,p70,Interleukin-12 | |
Unconjugated | |
His Tag and Flag Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |