missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IGSF1 Partial ORF (NP_001546, 220 a.a. - 310 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00003547-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16126721

  • 574.00 CHF / 25 Mikrogramm
Verfügbar ab: 15-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Members of the immunoglobulin (Ig) superfamily (see MIM 147100), which includes IGSF1, have a variety of functions, but all appear to play a role in cell recognition and the regulation of cell behavior.[supplied by OMIM]

Sequence: VVAGLYPKPTLTAHPGPIMAPGESLNLRCQGPIYGMTFALMRVEDLEKSFYHKKTIKNEANFFFQSLKIQDTGHYLCFYYDASYRGSLLSD
Spezifikation

Spezifikation

NP_001546
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VVAGLYPKPTLTAHPGPIMAPGESLNLRCQGPIYGMTFALMRVEDLEKSFYHKKTIKNEANFFFQSLKIQDTGHYLCFYYDASYRGSLLSD
RUO
IGSF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3547
IGSF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IGCD1/IGDC1/INHBP/KIAA0364/MGC75490/PGSF2
IGSF1
Recombinant
wheat germ expression system
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote