missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgM Antibody, R&D Systems™

Rabbit Monoclonal Antibody
264.00 CHF - 546.00 CHF
Spezifikation
| Antigen | IgM |
|---|---|
| Klon | 2123A |
| Verdünnung | Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, ELISA |
| Anwendungen | Western Blot, ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18716739
![]() |
R&D Systems
MAB9435-SP |
25 μg |
264.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18783189
![]() |
R&D Systems
MAB9435-100 |
100 μg |
546.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Western Blot, Simple Western, ELISA.Spezifikation
| IgM | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 3507 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2123A | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Immunoglobulin M (IgM), mu chain from human plasma | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IgM in Western blots. |
For Research Use Only (RUO)
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts