missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG Fc Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marke: R&D Systems G-102-C
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgG Polyclonal specifically detects IgG in Human samples. It is validated for .
Spezifikation
| IgG | |
| LYOPH | |
| Hidden | |
| Immunoglobulin G, ImmunoglobulinG | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 3500 | |
| Reconstitute with sterile PBS. If 0.5 mL of PBS is used, the antibody concentration will be 0.2 mg/mL | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with No Preservative | |
| IGHG1 | |
| Human IgG | |
| 0.1 mg | |
| Secondary | |
| This antibody reacts with heavy chains on human IgG, but not with light chains on immunoglobulins. No reactivity is observed to non-immunoglobulin human serum proteins by IEP | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur