missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human IGFBP-4 Protein, His Tag
378.31 CHF - 2799.42 CHF
Spezifikation
Zugriffsnummer | NP_001543.2 |
---|---|
Molekulargewicht | 27.9 kDa |
Quelle | Human IGFBP-4, His Tag (IG4-H52E4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Glu 258 (Accession # P22692-1). |
Immunogen | Asp 22 - Glu 258 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15837007
|
ACROBiosystems
IG4-H52E4-50UG |
50 μg |
378.31 CHF
50 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827007
|
ACROBiosystems
IG4-H52E4-1MG |
1 mg |
2799.42 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Insulin-like growth factor-binding protein 4 (IGFBP-4) is also known as IGF-binding protein 4, which contains an IGFBP domain and a thyroglobulin type-I domain. IGFBP-4 binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors. IGFBP-4 is a unique protein and it consistently inhibits several cancer cells in vivo and in vitro. Its inhibitory action has been shown in vivo in prostate and colon. IGFBP-4 is secreted by all colon cancer cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 33-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001543.2 | |
Human IGFBP-4, His Tag (IG4-H52E4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Glu 258 (Accession # P22692-1). | |
-20°C | |
IGFBP-4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
27.9 kDa | |
Asp 22 - Glu 258 | |
IGFBP4,IBP-4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |