missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IGF2 ELISA Kit

Artikelnummer. 31369620
Ansicht ändern
Click to view available options
Menge:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31369620 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31369620 Lieferant Invitrogen™ Lieferanten-Nr. EH555RB
 Eingeschränkter Artikel
Verfügbar ab: 04-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

Human IGF2 ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human IGF-2 in serum, plasma, and cell culture supernatants.

The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P01344
Assay-Bereich 4.1-1000 ng/mL
Assay-Empfindlichkeit: 4.1 ng/mL
Konjugat HRP
Immunassay-Kit-Format Sandwich ELISA Kit
Produkttyp ELISA
Probentyp Plasma, Serum, Supernatant
Zielspezies Human
Zur Verwendung mit (Geräte) Absorbance Microplate Reader
Gen-Alias AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factorAL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor;
Gen-ID (Entrez) 3481
Gensymbol IGF2
Interassay-VK 12%
Intraassay-VK 10%
Kitinhalt Pre-coated microwell plate, Standard, Biotinylated Detection Antibody, Streptavidin-HRP, Sample Diluent, Wash Buffer Concentrate, Assay Buffer concentrate, Stop Solution, Substrate Solution, Adhesive Plate Covers
Marker oder Farbstoff Biotin
Menge 96 Tests
Kennzeichnung RUO
Versandbedingung Wet Ice
Lagerungsbedingungen -20°C
Ziel IGF2
Prüfzeit 1 hr 20 min
Gesamte Assay-Zeit 4 hr 45 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.