missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HSPH1 Full-length ORF (NP_006635.2, 1 a.a. - 858 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_006635.2 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 10808 |
Molekulargewicht | 123.3kDa |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
16145732
|
Abnova™
H00010808-P02.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16135732
|
Abnova™
H00010808-P02.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
Sequence: MSVVGLDVGSQSCYIAVARAGGIETIANEFSDRCTPSVISFGSKNRTIGVAAKNQQITHANNTVSNFKRFHGRAFNDPFIQKEKENLSYDLVPLKNGGVGIKVMYMGEEHLFSVEQITAMLLTKLKETAENSLKKPVTDCVISVPSFFTDAERRSVLDAAQIVGLNCLRLMNDMTAVALNYGIYKQDLPSLDEKPRIVVFVDMGHSAFQVSACAFNKGKLKVLGTAFDPFLGGKNFDEKLVEHFCAEFKTKYKLDAKSKIRALLRLYQECEKLKKLMSSNSTDLPLNIECFMNDKDVSGKMNRSQFEELCAELLQKIEVPLYSLLEQTHLKVEDVSAVEIVGGATRIPAVKERIAKFFGKDISTTLNADEAVARGCALQCAILSPAFKVREFSVTDAVPFPISLIWNHDSEDTEGVHEVFSRNHAAPFSKVLTFLRRGPFELEAFYSDPQGVPYPEAKIGRFVVQNVSAQKDGEKSRVKVKVRVNTHGIFTISTASMVEKVPTEENEMSSEADMECLNQRPPENPDTDKNVQQDNSEAGTQPQVQTDAQQTSQSPPSPELTSEENKIPDADKANEKKVDQPPEAKKPKIKVVNVELPIEANLVWQLGKDLLNMYIETEGKMIMQDKLEKERNDAKNAVEEYVYEFRDKLCGPYEKFICEQDHQNFLRLLTETEDWLYEEGEDQAKQAYVDKLEELMKIGTPVKVRFQEAEERPKMFEELGQRLQHYAKIAADFRNKDEKYNHIDESEMKKVEKSVNEVMEWMNNVMNAQAKKSLDQDPVVRAQEIKTKIKELNNTCEPVVTQPKPKIESPKLERTPNGPNIDKKEEDLEDKNNFGAEPPHQNGECYPNEKNSVNMDLDSpecifications
NP_006635.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
123.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSVVGLDVGSQSCYIAVARAGGIETIANEFSDRCTPSVISFGSKNRTIGVAAKNQQITHANNTVSNFKRFHGRAFNDPFIQKEKENLSYDLVPLKNGGVGIKVMYMGEEHLFSVEQITAMLLTKLKETAENSLKKPVTDCVISVPSFFTDAERRSVLDAAQIVGLNCLRLMNDMTAVALNYGIYKQDLPSLDEKPRIVVFVDMGHSAFQVSACAFNKGKLKVLGTAFDPFLGGKNFDEKLVEHFCAEFKTKYKLDAKSKIRALLRLYQECEKLKKLMSSNSTDLPLNIECFMNDKDVSGKMNRSQFEELCAELLQKIEVPLYSLLEQTHLKVEDVSAVEIVGGATRIPAVKERIAKFFGKDISTTLNADEAVARGCALQCAILSPAFKVREFSVTDAVPFPISLIWNHDSEDTEGVHEVFSRNHAAPFSKVLTFLRRGPFELEAFYSDPQGVPYPEAKIGRFVVQNVSAQKDGEKSRVKVKVRVNTHGIFTISTASMVEKVPTEENEMSSEADMECLNQRPPENPDTDKNVQQDNSEAGTQPQVQTDAQQTSQSPPSPELTSEENKIPDADKANEKKVDQPPEAKKPKIKVVNVELPIEANLVWQLGKDLLNMYIETEGKMIMQDKLEKERNDAKNAVEEYVYEFRDKLCGPYEKFICEQDHQNFLRLLTETEDWLYEEGEDQAKQAYVDKLEELMKIGTPVKVRFQEAEERPKMFEELGQRLQHYAKIAADFRNKDEKYNHIDESEMKKVEKSVNEVMEWMNNVMNAQAKKSLDQDPVVRAQEIKTKIKELNNTCEPVVTQPKPKIESPKLERTPNGPNIDKKEEDLEDKNNFGAEPPHQNGECYPNEKNSVNMDLD | |
RUO | |
HSPH1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10808 | |
HSPH1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686M05240/HSP105/HSP105A/HSP105B/KIAA0201/NY-CO-25 | |
HSPH1 | |
Recombinant | |
wheat germ expression system |