Learn More
Abnova™ Human HOXD3 Partial ORF (NP_008829, 334 a.a. - 431 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The protein encoded by this gene may play a role in the regulation of cell adhesion processes. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_008829 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 3232 |
Molekulargewicht | 36.52kDa |
Name | HOXD3 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 µg |
Immunogen | FEPHPMASNGGGFASANLQGSPVYVGGNFVESMAPASGPVFNLGHLSHPSSASVDYSCAAQIPGNHHHGPCDPHPTYTDLSAHHSSQGRLPEAPKLTH |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.