Learn More
Abnova™ Human HOOK1 Full-length ORF (NP_056972.1, 1 a.a. - 728 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_056972.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 51361 |
Molekulargewicht | 111kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16129942
|
Abnova™
H00051361-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16119942
|
Abnova™
H00051361-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the hook family of coiled-coil proteins, which bind to microtubules and organelles through their N- and C-terminal domains, respectively. The encoded protein localizes to discrete punctuate subcellular structures, and interacts with several members of the Rab GTPase family involved in endocytosis. It is thought to link endocytic membrane trafficking to the microtubule cytoskeleton. Several alternatively spliced transcript variants have been identified, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MEETQPPPQPKLPLCDSLMIWLQTFNTASPCQDVKQLTSGVAMAQVLHQIDAAWFNESWLSRIKEDVGDNWRIKASNVKKVLQGIMSYYHEFLGQQISEALIPDLNQITECSDPVELGRLLQLILGCAINCEKKQEHIQNIMTLEESVQHVVMTAIQELMSKEILSSPPNDAVGELEQQLKRALEELQEALAEKEELRQRCEELDMQVTTLQDEKNSLVSENEMMNEKLDQLDGSFDDPNTVVAKKYFHAQLQLEQLQEENFRLEAAKDDYRVHCEELEKQLIEFQHRNDELTSLAEETRALKDEIDVLRATSDKANKLESTVEIYRQKLQDLNDLRKQVKTLQETNMMYMHNTVSLEEELKKANAARTQLETYKRQVQDLHVKLSSESKRADTLAFEMKRLEEKHEALLKEKERLIEQRDTLKETNEELRCSQVQQDHLNQTDASATKSYENLAAEIMPVEYREVFIRLQHENKMLRLQQEGSENERIEELQEQLEQKHRKMNELETEQRLSKERIRELQQQIEDLQKSLQEQGSKSEGESSSKLKQKLEAHMEKLTEVHEELQKKQELIEDLQPDINQNVQKINELEAALQKKDEDMKAMEERYKMYLEKARNVIKTLDPKLNPASAEIMLLRKQLAEKERRIEILESECKVAKFRDYEEKLIVSAWYNKSLAFQKLGMESRLVSGGGACSDTGACTPARSFLAQQRHITNTRRNLSVKVPATTSDSpezifikation
NP_056972.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
111kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEETQPPPQPKLPLCDSLMIWLQTFNTASPCQDVKQLTSGVAMAQVLHQIDAAWFNESWLSRIKEDVGDNWRIKASNVKKVLQGIMSYYHEFLGQQISEALIPDLNQITECSDPVELGRLLQLILGCAINCEKKQEHIQNIMTLEESVQHVVMTAIQELMSKEILSSPPNDAVGELEQQLKRALEELQEALAEKEELRQRCEELDMQVTTLQDEKNSLVSENEMMNEKLDQLDGSFDDPNTVVAKKYFHAQLQLEQLQEENFRLEAAKDDYRVHCEELEKQLIEFQHRNDELTSLAEETRALKDEIDVLRATSDKANKLESTVEIYRQKLQDLNDLRKQVKTLQETNMMYMHNTVSLEEELKKANAARTQLETYKRQVQDLHVKLSSESKRADTLAFEMKRLEEKHEALLKEKERLIEQRDTLKETNEELRCSQVQQDHLNQTDASATKSYENLAAEIMPVEYREVFIRLQHENKMLRLQQEGSENERIEELQEQLEQKHRKMNELETEQRLSKERIRELQQQIEDLQKSLQEQGSKSEGESSSKLKQKLEAHMEKLTEVHEELQKKQELIEDLQPDINQNVQKINELEAALQKKDEDMKAMEERYKMYLEKARNVIKTLDPKLNPASAEIMLLRKQLAEKERRIEILESECKVAKFRDYEEKLIVSAWYNKSLAFQKLGMESRLVSGGGACSDTGACTPARSFLAQQRHITNTRRNLSVKVPATTSD | |
RUO | |
HOOK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51361 | |
HOOK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HK1/MGC10642 | |
HOOK1 | |
Recombinant | |
wheat germ expression system |