Learn More
Abnova™ Human HNRNPG-T Partial ORF (NP_055284, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene belongs to the HNRPG subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two RRM domains that bind RNAs. This gene is intronless and is thought to be derived from a processed retroposon. However, unlike many retroposon-derived genes, this gene is not a pseudogene. The encoded protein has similarity to HNRPG and RBMY proteins and it is suggested to replace HNRPG protein function during meiotic prophase or act as a germ cell-specific splicing regulator. It primarily localizes to the nuclei of meiotic spermatocytes. This gene is a candidate for autosomal male infertility. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_055284 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 27288 |
Molekulargewicht | 35.64kDa |
Name | HNRNPG-T (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | MVEADRPGKLFIGGLNLETDEKALEAEFGKYGRIVEVLLMKDRETNKSRGFAFVTFESPADAKAAARDMNGKSLDGKAIKVAQATKPAFE |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.