missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HNF4G Partial ORF (NP_004124.3, 310 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003174-Q01.10ug
Dettagli aggiuntivi : Gewicht : 0.00010kg
Descrizione
Sequence: KLFGMVKIDNLLQEMLLGGASNDGSHLHHPMHPHLSQDPLTGQTILLGPMSTLVHADQISTPETPLPSPPQGSGQEQYKIAANQASVISHQHLSKQKQSpecifica
NP_004124.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KLFGMVKIDNLLQEMLLGGASNDGSHLHHPMHPHLSQDPLTGQTILLGPMSTLVHADQISTPETPLPSPPQGSGQEQYKIAANQASVISHQHLSKQKQ | |
RUO | |
HNF4G | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3174 | |
HNF4G (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NR2A2/NR2A3 | |
HNF4G | |
Recombinant | |
wheat germ expression system |