missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HIF-2 alpha/EPAS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB2886
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Western Blot.
Spezifikation
| HIF-2 alpha/EPAS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q99814 | |
| EPAS1 | |
| E. coli-derived recombinant human HIF-2 alpha/EPAS1 Ser543-Thr870 Accession # Q99814 | |
| 100 μg | |
| Primary | |
| Detects human HIF-2 alpha/EPAS1 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 318404 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2034 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur