missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HEATR2 Full-length ORF (AAH10850.1, 1 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00054919-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16150893

  • 458.00 CHF / 25 Mikrogramm


Verfügbar ab
03-07-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MRLKLFSILSTVLLRATDTINSQGQFPSYLETVTKDILAPNLQWHAGRTAAAIRTAAVSCLWALTSSEVLSAEQIRDVQETLMPQVLTTLEEDSKMTRLISCRIINTFLKTSGGMTDPEKLIKIYPELLKRLDDVSNDVRMAAASTLVTWLQCVKGANAKSYYQSSVQYLYRELLVHLDDPERAIQDAILDVPGSPSPSAMIGSFLRPPHPWRTVSQLNLFSS
Spezifikation

Spezifikation

AAH10850.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20397|FLJ25564|FLJ31671|FLJ39381
HEATR2
Yes
wheat germ expression system
Liquid
54919
HEATR2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLKLFSILSTVLLRATDTINSQGQFPSYLETVTKDILAPNLQWHAGRTAAAIRTAAVSCLWALTSSEVLSAEQIRDVQETLMPQVLTTLEEDSKMTRLISCRIINTFLKTSGGMTDPEKLIKIYPELLKRLDDVSNDVRMAAASTLVTWLQCVKGANAKSYYQSSVQYLYRELLVHLDDPERAIQDAILDVPGSPSPSAMIGSFLRPPHPWRTVSQLNLFSS
RUO
HEATR2
Wheat Germ (in vitro)
GST
Videos
Dokumentation