Learn More
Abnova™ Human H6PD Partial ORF (NP_004276, 401 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009563-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
There are 2 forms of glucose-6-phosphate dehydrogenase. G form is X-linked and H form, encoded by this gene, is autosomally linked. This H form shows activity with other hexose-6-phosphates, especially galactose-6-phosphate, whereas the G form is specific for glucose-6-phosphate. Both forms are present in most tissues, but H form is not found in red cells. [provided by RefSeq]
Sequence: HIGHGDLGSPAVLVSRNLFRPSLPSSWKEMEGPPGLRLFGSPLSDYYAYSPVRERDAHSVLLSHIFHGRKNFFITTENLLASWNFWTPLLESLAHKAPRLSpezifikation
NP_004276 | |
Liquid | |
9563 | |
H6PD (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A01246/G6PDH/GDH/MGC87643 | |
H6PD | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HIGHGDLGSPAVLVSRNLFRPSLPSSWKEMEGPPGLRLFGSPLSDYYAYSPVRERDAHSVLLSHIFHGRKNFFITTENLLASWNFWTPLLESLAHKAPRL | |
RUO | |
H6PD | |
Wheat Germ (in vitro) | |
GST |