missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GTF2H3 Partial ORF (NP_001507, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002967-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MVSDEDELNLLVIVVDANPIWWGKQALKESQFTLSKCIDAVMVLGNSHLFMNRSNKLAVIASHIQESRFLYPGKNGRLGDFFGDPGNPPEFNPSGSKDGKYELLTSANEVSpezifikation
NP_001507 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVSDEDELNLLVIVVDANPIWWGKQALKESQFTLSKCIDAVMVLGNSHLFMNRSNKLAVIASHIQESRFLYPGKNGRLGDFFGDPGNPPEFNPSGSKDGKYELLTSANEV | |
RUO | |
GTF2H3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2967 | |
GTF2H3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BTF2/TFB4/TFIIH | |
GTF2H3 | |
Recombinant | |
wheat germ expression system |