Learn More
ACROBiosystems Human Growth Hormone R (GHR) Protein, Fc Tag
Marke: ACROBiosystems GHR-H5257-50UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Growth hormone receptor (GHR) is also known as somatotropin receptor, growth hormone-binding protein (GHBR), which belongs to the type I cytokine receptor family or Type 1 subfamily. GHR contains one fibronectin type-III domain. GHR / GHBR is expressed in various tissues with high expression in liver and skeletal muscle. The soluble form (GHBP) is produced by phorbol ester-promoted proteolytic cleavage at the cell surface (shedding) by ADAM17/TACE. GHR is receptor for pituitary gland growth hormone involved in regulating postnatal body growth. On ligand binding, couples to the JAK2/STAT5 pathway. The soluble form (GHBP) acts as a reservoir of growth hormone in plasma and may be a modulator/inhibitor of GH signaling.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. As a result of glycosylation, the protein migrates as 66-85 kDa under reducing (R) condition, and 135-160 kDa under non-reducing (NR) condition (SDS-PSpezifikation
P10912 | |
50 μg | |
Ala 27 - Tyr 264 | |
GHR,GHBP,GH receptor | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
54.3 kDa | |
Human GHR, Fc Tag (GHR-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Tyr 264 (Accession # P10912). | |
-20°C | |
Growth Hormone R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |