missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human GPA33 / A33 Protein, His Tag
327.86 CHF - 2383.29 CHF
Spezifikation
Zugriffsnummer | AAH74830 |
---|---|
Molekulargewicht | 24.4 kDa |
Quelle | Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # AAH74830). |
Immunogen | Ile 22 - Val 235 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15816877
|
ACROBiosystems
GP3-H5224-1MG |
1 mg |
2383.29 CHF
1 Milligramm |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826877
|
ACROBiosystems
GP3-H5224-50UG |
50 μg |
327.86 CHF
50 Mikrogramm |
Verfügbar ab: 30-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Glycoprotein A33 (GPA33) is also known as Cell surface A33 antigen, is a single-pass type I membrane protein which is expressed in normal gastrointestinal epithelium and in 95% of colon cancers. GPA33 The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily, which contains. GPA33 may play a role in cell-cell recognition and signaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH74830 | |
Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # AAH74830). | |
-20°C | |
GPA33 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
24.4 kDa | |
Ile 22 - Val 235 | |
GPA33,A33 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>96% as determined by SDS-PAGE |